5-Bromo-4-fluoro-2-methylbenzamide ≥97%

Supplier: AOBChem USA

121599-1G 121599-25G 121599-500MG 121599-5G
76924-704EA 319.34 USD
76924-704 76924-706 76924-708 76924-710
5-Bromo-4-fluoro-2-methylbenzamide ≥97%
5-Bromo-4-fluoro-2-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
CAS Number: 2092621-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR