4-Isopropoxy-3-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Isopropoxy-3-methylbenzeneboronic acid

12129-25G 12129-5G
76924-682EA 431.79 USD
76924-682 76924-684
4-Isopropoxy-3-methylphenylboronic acid ≥97%
4-Isopropoxy-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD06659871
CAS Number: 850568-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR