5-Ethoxy-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Dihydroxy(5-ethoxy-2-fluorophenyl)borane, (5-Ethoxy-2-fluorophenyl)boronic acid, 5-Ethoxy-2-fluorobenzeneboronic acid, (5-Ethoxy-2-fluorophenyl)metaboric acid

11938-25G 11938-5G
76924-640EA 240.83 USD
76924-640 76924-642
5-Ethoxy-2-fluorophenylboronic acid ≥97%
5-Ethoxy-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD05664321
CAS Number: 900174-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR