4-(Ethoxycarbonyl)-2-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Ethoxycarbonyl-2-nitrophenylboronic acid, 4-Ethoxycarbonyl-2-nitrobenzeneboronic acid

11929-25G 11929-5G
76924-634EA 198.39 USD
76924-634 76924-636
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid ≥97%
4-(Ethoxycarbonyl)-2-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₆
Storage Temperature: Ambient
MDL Number: MFCD02179460
CAS Number: 5785-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR