4-Chloro-3-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

11506-25G 11506-5G
76924-598EA 198.39 USD
76924-598 76924-600
4-Chloro-3-fluoropyridine ≥97%
4-Chloro-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFN
Storage Temperature: Refrigerator
MDL Number: MFCD03453233
CAS Number: 2546-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR