5-Chloro-2-fluoro-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-fluoropyridine-3-boronic acid, (5-Chloro-2-fluoropyridin-3-yl)boronic acid, 5-Chloro-2-fluoro-3-pyridylboronic acid

11466-25G 11466-5G
76924-582EA 492.26 USD
76924-582 76924-584
5-Chloro-2-fluoro-3-pyridineboronic acid ≥97%
5-Chloro-2-fluoro-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BClFNO₂
MW: 175.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972408
CAS Number: 937595-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR