5-Chloro-2,4-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2,4,-difluorophenylboronic acid, (5-Chloro-2,4-difluorophenyl)boronic acid, 5-Chloro-2,4-difluorobenzeneboronic acid

11451-1G 11451-25G 11451-500MG 11451-5G
76924-570EA 124.13 USD
76924-570 76924-572 76924-574 76924-576
5-Chloro-2,4-difluorophenylboronic acid ≥97%
5-Chloro-2,4-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BClF₂O₂
MW: 192.3600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038751
CAS Number: 911645-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR