4-Cyano-2,6-difluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

11264-1G 11264-25G 11264-500MG 11264-5G
76924-522EA 212.18 USD
76924-522 76924-524 76924-526 76924-528
4-Cyano-2,6-difluorophenylboronic acid ≥95%
4-Cyano-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₄BF₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03095140
CAS Number: 2199440-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR