N,N-Dibenzyl-4-bromoaniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-N,N-dibenzylaniline

11212-25G 11212-5G
76924-502EA 488.02 USD
76924-502 76924-504
N,N-Dibenzyl-4-bromoaniline ≥97%
N,N-Dibenzyl-4-bromoaniline
Formula: C₂₀H₁₈BrN
MW: 352.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037473
CAS Number: 65145-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR