4-Butoxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-N-Butoxy-2-methylphenylboronic acid

11101-25G 11101-5G
76924-490EA 453.01 USD
76924-490 76924-492
4-Butoxy-2-methylphenylboronic acid ≥97%
4-Butoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD05664325
CAS Number: 845551-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR