4-BUTOXY-3,5-DIMETHYLPHENYLBORONIC ACID ≥97%

Supplier: AOBChem USA

Warning

10947-1G 10947-25G 10947-5G
76924-454EA 62.6 USD
76924-454 76924-456 76924-458
4-BUTOXY-3,5-DIMETHYLPHENYLBORONIC ACID ≥97%
4-BUTOXY-3,5-DIMETHYLPHENYLBORONIC ACID

Precaución: for Research Use Only

Formula: C₁₂H₁₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD05865176
CAS Number: 845551-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR