4-Bromo-2-fluoronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

108898-1G 108898-25G 108898-500MG 108898-5G
76924-438EA 367.08 USD
76924-438 76924-440 76924-442 76924-444
4-Bromo-2-fluoronicotinic acid ≥97%
4-Bromo-2-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MW: 219.996 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18258640
CAS Number: 1256790-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR