5-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10566-25G 10566-5G
76924-382EA 204.76 USD
76924-382 76924-384
5-Fluoro-2-(trifluoromethyl)phenylboronic acid ≥97%
5-Fluoro-2-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₂
MW: 207.9180 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656275
CAS Number: 928053-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR