4-(Ethoxycarbonyl)-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10539-25G 10539-5G
76924-374EA 300.24 USD
76924-374 76924-376
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid ≥97%
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD06656274
CAS Number: 874288-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR