5-Fluoro-(2-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoro-2-methoxycarboxyphenylboronic acid, 5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid, 2-Methoxycarbonyl-5-fluorophenylboronic acid

10491-1G 10491-25G 10491-5G
76924-356EA 64.72 USD
76924-356 76924-358 76924-360
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid ≥97%
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095367
CAS Number: 850568-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR