2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine-2-carboxylic acid methylamide, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

10455-25G 10455-5G
76924-326EA 598.35 USD
76924-326 76924-328
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester ≥97% research grade
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester
Formula: C₁₃H₁₉BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08458483
CAS Number: 945863-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR