4-Bromoacetyl-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

10354-1G 10354-25G 10354-5G
76924-290EA 62.6 USD
76924-290 76924-292 76924-294
4-Bromoacetyl-3-fluorophenylboronic acid ≥97%
4-Bromoacetyl-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BBrFO₃
MW: 260.853 g/mol
MDL Number: MFCD06796423
CAS Number: 481725-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR