4-Carboxy-2-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-2-nitrobenzeneboronic acid

10222-25G 10222-5G
76924-270EA 268.41 USD
76924-270 76924-272
4-Carboxy-2-nitrophenylboronic acid ≥97%
4-Carboxy-2-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BNO₆
Storage Temperature: Ambient
MDL Number: MFCD00448780
CAS Number: 85107-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR