4-Chloro-3-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-nitrobenzeneboronic acid

10187-25G 10187-5G
76924-256EA 198.39 USD
76924-256 76924-258
4-Chloro-3-nitrophenylboronic acid ≥97%
4-Chloro-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD02258950
CAS Number: 151169-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR