(5-Bromopicolinoyl)piperidine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-(piperidin-1-ylcarbonyl)pyridine, (5-Bromopyridin-2-yl)(piperidin-1-yl)methanone

10149-1G 10149-25G 10149-5G
76924-224EA 81.69 USD
76924-224 76924-226 76924-228
(5-Bromopicolinoyl)piperidine ≥97% research grade
(5-Bromopicolinoyl)piperidine
Formula: C₁₁H₁₃BrN₂O
MW: 269.1377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458472
CAS Number: 934000-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR