5-Bromo-N-methylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-N-methylpyridine-2-carboxamide

10139-25G 10139-5G
76924-204EA 495.45 USD
76924-204 76924-206
5-Bromo-N-methylpicolinamide ≥97%
5-Bromo-N-methylpicolinamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07357421
CAS Number: 845305-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR