5-Chloro-2-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10066-25G 10066-5G
76924-198EA 293.87 USD
76924-198 76924-200
5-Chloro-2-formylphenylboronic acid ≥97%
5-Chloro-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
MW: 184.3900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425955
CAS Number: 870238-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR