2-Bromo-3,6-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

95311-1G 95311-25G 95311-500MG 95311-5G
76906-124EA 344.8 USD
76906-124 76906-126 76906-128 76906-130
2-Bromo-3,6-dimethoxybenzaldehyde ≥97%
2-Bromo-3,6-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00457771
CAS Number: 85432-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR