2-Fluoro-3-(methylthio)-6-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

93384-1G 93384-25G 93384-500MG 93384-5G
76906-112EA 136.86 USD
76906-112 76906-114 76906-116 76906-118
2-Fluoro-3-(methylthio)-6-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-3-(methylthio)-6-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆F₄OS
MW: 238.202 g/mol
Storage Temperature: Ambient
CAS Number: 2921869-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR