2-Fluoro-3-(methylthio)-6-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

92425-1G 92425-25G 92425-500MG 92425-5G
76906-100EA 123.07 USD
76906-100 76906-102 76906-104 76906-106
2-Fluoro-3-(methylthio)-6-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-(methylthio)-6-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂S
MW: 254.201 g/mol
Storage Temperature: Ambient
CAS Number: 2921836-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR