2-Chloro-4-hydroxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

92237-1G 92237-500MG 92237-5G
76906-088EA 814.78 USD
76906-088 76906-090 76906-092
2-Chloro-4-hydroxy-3-methylbenzoic acid ≥97%
2-Chloro-4-hydroxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
Storage Temperature: Ambient
CAS Number: 1824109-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR