2,5-Dibromo-4-fluorobenzaldehyde 95%

Supplier: AOBChem USA

Warning

88805-1G 88805-500MG 88805-5G 88805-25G
76906-050EA 458.31 USD
76906-050 76906-052 76906-054 77660-494
2,5-Dibromo-4-fluorobenzaldehyde 95%
2,5-Dibromo-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Br₂FO
MW: 281.905 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847512
CAS Number: 1379332-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR