2-Bromo-4-methyl-5-(trifluoromethyl)aniline ≥97%

Supplier: AOBChem USA

Warning

87169-1G 87169-25G 87169-500MG 87169-5G
76906-034EA 387.23 USD
76906-034 76906-036 76906-038 76906-040
2-Bromo-4-methyl-5-(trifluoromethyl)aniline ≥97%
2-Bromo-4-methyl-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₈H₇BrF₃N
MW: 254.047 g/mol
Storage Temperature: Ambient
CAS Number: 1260776-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR