2-Bromo-5-(cyclopropylmethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

86112-1G 86112-25G 86112-5G
76906-022EA 76.39 USD
76906-022 76906-024 76906-026
2-Bromo-5-(cyclopropylmethoxy)pyridine ≥97%
2-Bromo-5-(cyclopropylmethoxy)pyridine
Formula: C₉H₁₀BrNO
MW: 228.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22690535
CAS Number: 1177269-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR