1-Chloro-2-fluoro-6-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

83854-25G 83854-5G
76906-014EA 453.01 USD
76906-014 76906-016
1-Chloro-2-fluoro-6-iodobenzene ≥97%
1-Chloro-2-fluoro-6-iodobenzene
Formula: C₆H₃ClFI
Storage Temperature: Ambient
MDL Number: MFCD11100163
CAS Number: 348642-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR