2-Ethoxy-3-ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

83444-1G 83444-25G 83444-500MG 83444-5G
76905-998EA 173.99 USD
76905-998 76906-000 76906-002 76906-004
2-Ethoxy-3-ethylbenzoic acid ≥97%
2-Ethoxy-3-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
CAS Number: 2007434-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR