2-Chloro-5-iodo-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

82813-1G 82813-25G 82813-500MG 82813-5G
76905-982EA 196.27 USD
76905-982 76905-984 76905-986 76905-988
2-Chloro-5-iodo-3-methoxypyridine ≥97%
2-Chloro-5-iodo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClINO
Storage Temperature: Ambient
CAS Number: 1352881-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR