2-Methoxy-5-(methoxymethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

79725-1G 79725-25G 79725-500MG 79725-5G
76905-930EA 163.38 USD
76905-930 76905-932 76905-934 76905-936
2-Methoxy-5-(methoxymethoxy)benzaldehyde ≥97%
2-Methoxy-5-(methoxymethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
Storage Temperature: Refrigerator
CAS Number: 142677-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR