2-Chloro-3-hydroxyisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

78130-1G 78130-25G 78130-500MG 78130-5G
76905-916EA 143.23 USD
76905-916 76905-918 76905-920 76905-922
2-Chloro-3-hydroxyisonicotinic acid ≥97%
2-Chloro-3-hydroxyisonicotinic acid
Formula: C₆H₄ClNO₃
Storage Temperature: Ambient
CAS Number: 185423-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR