2-Chloro-5-fluoro-3-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

77710-1G 77710-500MG 77710-5G 77710-25G
76905-904EA 1088.49 USD
76905-904 76905-906 76905-908 77660-332
2-Chloro-5-fluoro-3-isopropoxybenzoic acid ≥97%
2-Chloro-5-fluoro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
MW: 232.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767204
CAS Number: 2918901-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR