2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

76687-1G 76687-25G 76687-500MG 76687-5G
76905-892EA 439.22 USD
76905-892 76905-894 76905-896 76905-898
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid ≥97%
2-Bromo-6-fluoro-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31759997
CAS Number: 1800423-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR