2-Bromo-3,4-dichloro-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

72942-1G 72942-25G 72942-500MG 72942-5G
76905-802EA 132.62 USD
76905-802 76905-804 76905-806 76905-808
2-Bromo-3,4-dichloro-6-methylaniline ≥97%
2-Bromo-3,4-dichloro-6-methylaniline

Precaución: for Research Use Only

Formula: C₇H₆BrCl₂N
Storage Temperature: Ambient
CAS Number: 1845713-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR