2-Bromo-5-iodo-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

72330-1G 72330-25G 72330-500MG 72330-5G
76905-786EA 363.89 USD
76905-786 76905-788 76905-790 76905-792
2-Bromo-5-iodo-4-methylphenol ≥97%
2-Bromo-5-iodo-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.93 g/mol
Storage Temperature: Ambient
CAS Number: 2090127-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR