2-Chloro-1-fluoro-4-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

71519-25G 71519-5G
76905-774EA 587.74 USD
76905-774 76905-776
2-Chloro-1-fluoro-4-isobutoxybenzene ≥97%
2-Chloro-1-fluoro-4-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 1852162-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR