2,5-Dibromo-4-fluoroaniline ≥97% research grade

Supplier: AOBChem USA

Warning

69452-25G 69452-5G
76905-766EA 418 USD
76905-766 76905-768
2,5-Dibromo-4-fluoroaniline ≥97% research grade
2,5-Dibromo-4-fluoroaniline
Formula: C₆H₄Br₂FN
MW: 268.909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089317
CAS Number: 172377-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR