2-Chloro-N,N-dimethylnicotinamide ≥97%

Supplier: AOBChem USA

Warning

68854-25G 68854-5G
76905-740EA 595.17 USD
76905-740 76905-742
2-Chloro-N,N-dimethylnicotinamide ≥97%
2-Chloro-N,N-dimethylnicotinamide
Formula: C₈H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD08593346
CAS Number: 52943-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR