2-Iodo-1,4-dimethoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

68308-1G 68308-25G 68308-5G
76905-730EA 82.76 USD
76905-730 76905-732 76905-734
2-Iodo-1,4-dimethoxybenzene ≥97% research grade
2-Iodo-1,4-dimethoxybenzene
Formula: C₈H₉IO₂
MW: 264.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09032417
CAS Number: 25245-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR