2-Bromo-1-chloro-3,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

66740-1G 66740-25G 66740-500MG 66740-5G
76905-716EA 356.47 USD
76905-716 76905-718 76905-720 76905-722
2-Bromo-1-chloro-3,4-difluorobenzene ≥97%
2-Bromo-1-chloro-3,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD12922585
CAS Number: 1208077-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR