2-Fluoro-N,4-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

65335-1G 65335-25G 65335-500MG 65335-5G
76905-700EA 138.98 USD
76905-700 76905-702 76905-704 76905-706
2-Fluoro-N,4-dimethylbenzamide ≥97%
2-Fluoro-N,4-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO
Storage Temperature: Ambient
CAS Number: 1538873-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR