2-Bromo-5-chloro-4-fluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

65157-1G 65157-25G 65157-500MG 65157-5G
76905-684EA 292.81 USD
76905-684 76905-686 76905-688 76905-690
2-Bromo-5-chloro-4-fluorobenzoic acid ≥97% research grade
2-Bromo-5-chloro-4-fluorobenzoic acid
Formula: C₇H₃BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD24108577
CAS Number: 1519109-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR