2-Iodo-4-(trifluoromethoxy)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

64334-1G 64334-25G 64334-500MG 64334-5G
76905-656EA 397.84 USD
76905-656 76905-658 76905-660 76905-662
2-Iodo-4-(trifluoromethoxy)-1,1'-biphenyl ≥95%
2-Iodo-4-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₈F₃IO
MW: 364.1016 g/mol
Storage Temperature: Ambient
CAS Number: 2807455-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR