2-Chloro-4-(difluoromethoxy)-3-fluorobenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
63572-1G
63572-500MG
63572-5G
63572-25G
76905-626EA
1094.85
USD
76905-626
76905-628
76905-630
77656-746
2-Chloro-4-(difluoromethoxy)-3-fluorobenzoic acid ≥95%
2-Chloro-4-(difluoromethoxy)-3-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₄ClF₃O₃ MW: 240.5637 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34761429 CAS Number: 2385381-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...