2-Bromo-3-chloroanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-1-chloro-3-methoxybenzene

63275-25G 63275-5G
76905-616EA 410.57 USD
76905-616 76905-618
2-Bromo-3-chloroanisole ≥97%
2-Bromo-3-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390300
CAS Number: 174913-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR