2-Bromo-4-fluoro-1-(methoxymethoxy)-3-methylbenzene 95%

Supplier: AOBChem USA

Warning

62850-1G 62850-500MG 62850-5G 62850-25G
76905-570EA 678.98 USD
76905-570 76905-572 76905-574 77656-724
2-Bromo-4-fluoro-1-(methoxymethoxy)-3-methylbenzene 95%
2-Bromo-4-fluoro-1-(methoxymethoxy)-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.0769 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34761073
CAS Number: 2918951-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR