3-(4-Fluorobenzyloxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-((4-Fluorobenzyl)oxy)benzoic acid

62508-1G 62508-25G 62508-500MG 62508-5G
76905-550EA 130.5 USD
76905-550 76905-552 76905-554 76905-556
3-(4-Fluorobenzyloxy)benzoic acid ≥97%
3-(4-Fluorobenzyloxy)benzoic acid
Formula: C₁₄H₁₁FO₃
MW: 246.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04536523
CAS Number: 457-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR